missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD183 (CXCR3) Mouse anti-Human, PE-eFluor 610, Clone: CEW33D, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $487.08

Specifications

Antigen CD183 (CXCR3)
Clone CEW33D
Host Species Murine
Gene Alias C-X-C chemokine receptor type 3, CXC-R3, GPR9
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124790 Life Technologies
LS61183941
25 Tests Each for $196.68
501124791 Life Technologies
LS61183942
100 Tests Each for $487.08
Specifications

Description

The CEW33D monoclonal antibody reacts with human CD183. CD183, also known as CXCR3, is a G protein-coupled chemokine receptor that interacts with ligands CXCL9 (MIG), CXCL10 (IP-10), and CXCL11 (I-TAC). Strongly associated with type 1 immunity, CD183 is induced in naïve T cells upon activation and remains upregulated in T helper type (Th)1 cells, CD8 effector cells, NK cells and NKT cells. CD183-ligand interactions mediate infiltration of inflamed tissues in normal type 1 immune responses as well as in many inflammatory and autoimmune diseases. CD183 is also expressed on some B cells and plasmacytoid DC.
Description & Specifications

Specifications

CD183 (CXCR3)
Murine
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test
CEW33D
C-X-C chemokine receptor type 3, CXC-R3, GPR9
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer