Please login to your online account to display your discounted pricing

CD183 (CXCR3) Mouse anti-Human, PE-Cy7, Clone: CEW33D, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $535.05

Specifications

Antigen CD183 (CXCR3)
Clone CEW33D
Host Species Murine
Gene Alias C-X-C chemokine receptor type 3, CXC-R3, GPR9
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123349

 
life technologies
LS25183941
25 Tests Each for $216.05

501123350

 
life technologies
LS25183942
100 Tests Each for $535.05
Description & Specifications

Specifications

Antigen CD183 (CXCR3)
Clone CEW33D
Host Species Murine
Gene Alias C-X-C chemokine receptor type 3, CXC-R3, GPR9
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The CEW33D monoclonal antibody reacts with human CD183. CD183, also known as CXCR3, is a G protein-coupled chemokine receptor that interacts with ligands CXCL9 (MIG), CXCL10 (IP-10), and CXCL11 (I-TAC). Strongly associated with type 1 immunity, CD183 is induced in naïve T cells upon activation and remains upregulated in T helper type (Th)1 cells, CD8 effector cells, NK cells and NKT cells. CD183-ligand interactions mediate infiltration of inflamed tissues in normal type 1 immune responses as well as in many inflammatory and autoimmune diseases. CD183 is also expressed on some B cells and plasmacytoid DC.