missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD18 Rat anti-Mouse, PE, Clone: M18/2, eBioscience™

Rat Monoclonal Antibody

$104.28 - $249.48

Specifications

Antigen CD18
Clone M18/2
Host Species Rat
Gene Alias Integrin beta 2, ITGB2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010243 Life Technologies
LS12018181
50μg Each for $104.28
5010244 Life Technologies
LS12018182
100μg Each for $249.48
Specifications

Description

The M18/2 monoclonal antibody reacts with mouse CD18, the 90-95kDa integrin beta 2 glycoprotein. CD18 is the common β subunit that non-covalently associates with CD11a (alpha L), CD11b (alpha M) and CD11c (alpha X) to form LFA-1, Mac-1 and gp150/95. Heterodimers of CD18 and the a subunits are expressed broadly by all leukocytes with NK and T cells showing higher density of surface expression. Through the interactions of LFA-1, Mac-1 and gp150/95 with their counter-receptors, CD18 is involved in cellular adhesion and the immune response.
Description & Specifications

Specifications

CD18
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
M18/2
Integrin beta 2, ITGB2
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL