Please login to your online account to display your discounted pricing

CD18 Rat anti-Mouse, Clone: M18/2, eBioscience™

Rat Monoclonal Antibody

$71.05 - $332.05

Specifications

Antigen CD18
Clone M18/2
Host Species Rat
Gene Alias Integrin beta 2, ITGB2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012444

 
life technologies
LS14018181
50μg Each for $71.05

5012445

 
life technologies
LS14018182
100μg Each for $158.05

5012446

 
life technologies
LS14018185
500μg Each for $332.05
Description & Specifications

Specifications

Antigen CD18
Clone M18/2
Host Species Rat
Gene Alias Integrin beta 2, ITGB2
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The M18/2 monoclonal antibody reacts with mouse CD18, the 90-95kDa integrin beta 2 glycoprotein. CD18 is the common β subunit that non-covalently associates with CD11a (alpha L), CD11b (alpha M) and CD11c (alpha X) to form LFA-1, Mac-1 and gp150/95. Heterodimers of CD18 and the a subunits are expressed broadly by all leukocytes with NK and T cells showing higher density of surface expression. Through the interactions of LFA-1, Mac-1 and gp150/95 with their counter-receptors, CD18 is involved in cellular adhesion and the immune response.