Please login to your online account to display your discounted pricing

CD178 (Fas Ligand) Armenian Hamster anti-Mouse, FITC, Clone: MFL3, eBioscience™

Armenian Hamster Monoclonal Antibody

$129.05 - $292.90

Specifications

Antigen CD178 (Fas Ligand)
Clone MFL3
Host Species Armenian Hamster
Gene Alias CD95L, CD95 Ligand
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122150

 
life technologies
LS11591180
25μg Each for $129.05

5017737

 
life technologies
LS11591182
100μg Each for $292.90
Description & Specifications

Specifications

Antigen CD178 (Fas Ligand)
Clone MFL3
Host Species Armenian Hamster
Gene Alias CD95L, CD95 Ligand
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MFL3 monoclonal antibody reacts with mouse Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by mouse activated T cells. The interaction of FasL with its receptor CD95 induces Fas-mediated killing. It has been reported that the human FasL antigen is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. The degree of sensitivity for the mouse antigen to MMPs has not been reported.