missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD178 (Fas Ligand) Armenian Hamster anti-Mouse, Clone: MFL3, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$129.05 - $506.05

Specifications

Antigen CD178 (Fas Ligand)
Clone MFL3
Host Species Armenian Hamster
Gene Alias FasL, CD95L, CD95 Ligand
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122964 Life Technologies
LS16591185
500μg Each for $506.05
5014626 Life Technologies
LS16591181
50μg Each for $129.05
5014627 Life Technologies
LS16591182
100μg Each for $245.05
Specifications

Description

The MFL3 monoclonal antibody reacts with mouse Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by mouse activated T cells. The interaction of FasL with its receptor CD95 induces Fas-mediated killing. It has been reported that the human FasL antigen is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. The degree of sensitivity for the mouse antigen to MMPs has not been reported.
Description & Specifications

Specifications

CD178 (Fas Ligand)
Armenian Hamster
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
MFL3
FasL, CD95L, CD95 Ligand
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL