Please login to your online account to display your discounted pricing

CD178 (CD95 Ligand) Mouse anti-Human, PE, Clone: NOK-1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $390.05

Specifications

Antigen CD178 (CD95 Ligand)
Clone NOK-1
Host Species Mouse
Gene Alias Fas ligand, FasL
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122491

 
life technologies
LS12991941
25 tests Each for $172.55

5011290

 
life technologies
LS12991942
100 Tests Each for $390.05
Description & Specifications

Specifications

Antigen CD178 (CD95 Ligand)
Clone NOK-1
Host Species Mouse
Gene Alias Fas ligand, FasL
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The NOK-1 monoclonal antibody reacts with human Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by neutrophils, monocytes, and activated T cells and NK cells. The interaction of FasL with its receptor (CD95, Fas) induces Fas-mediated killing of lymphocytes. Human FasL is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. Therefore for optimal detection of surface FasL on activated peripheral blood cells, incubation of cells with an MMP inhibitor is recommended.