Please login to your online account to display your discounted pricing

CD178 (CD95 Ligand) Mouse anti-Human, Clone: NOK-1, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$158.05 - $477.05

Specifications

Antigen CD178 (CD95 Ligand)
Clone NOK-1
Host Species Mouse
Gene Alias CD95L, Fas ligand, FasL
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123013

 
affymetrix
16991985
500μg Each for $477.05

5014794

 
affymetrix
16991981
50μg Each for $158.05
Description & Specifications

Specifications

Antigen CD178 (CD95 Ligand)
Clone NOK-1
Host Species Mouse
Gene Alias CD95L, Fas ligand, FasL
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The NOK-1 monoclonal antibody reacts with human Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by neutrophils, monocytes, and activated T cells and NK cells. The interaction of FasL with its receptor (CD95, Fas) induces Fas-mediated killing of lymphocytes. Human FasL is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. Therefore for optimal detection of surface FasL on activated peripheral blood cells, incubation of cells with an MMP inhibitor is recommended.