Please login to your online account to display your discounted pricing

CD177 Mouse anti-Human, eFluor 660, Clone: MEM-166 (MEM166), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD177
Clone MEM-166 (MEM166)
Host Species Mouse
Gene Alias neutrophil-specific antigen, NB1, NB-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124316

 
life technologies
LS50177941
25 tests Each for $143.55

501124317

 
life technologies
LS50177942
100 tests Each for $288.55
Description & Specifications

Specifications

Antigen CD177
Clone MEM-166 (MEM166)
Host Species Mouse
Gene Alias neutrophil-specific antigen, NB1, NB-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.015μg)/test

The MEM-166 monoclonal antibody reacts with human CD177, a 58-64kDa GPI-anchored glycoprotein. The human CD177 locus codes for at least two alleles, NB1 and PRV-1, which differ by only 4 amino acids. CD177 is expressed on granulocytes, early erythroblasts, megakaryocytes, promyelocytes and myelocytes. CD177 mRNA levels are elevated in several different myeloproliferative disorders including polycythemia vera, and antibodies to CD177 are involved in neonatal alloimmune neutropenia, autoimmune drug induced neutropenia, and graft failure following marrow transplantation.