Please login to your online account to display your discounted pricing

CD172a (SIRP alpha) Mouse anti-Rat, APC, Clone: OX41, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen CD172a (SIRP alpha)
Clone OX41
Host Species Murine
Gene Alias SIRPa, SHPS1
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123093

 
life technologies
LS17172082
100μg Each for $375.55

501123092

 
life technologies
LS17172080
25μg Each for $172.55
Description & Specifications

Specifications

Antigen CD172a (SIRP alpha)
Clone OX41
Host Species Murine
Gene Alias SIRPa, SHPS1
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody OX41 reacts with rat CD172a, which is also known as signal-regulatory protein alpha (SIRPa). SIRP family members are transmembrane glycoproteins with 3 Ig-like extracellular domains and a cytoplasmic tail containing 2 immunocreceptor tyrosine-based inhibition motifs. CD172a is expressed by monocytes, macrophages, granulocytes, dendritic cells, and neurons. SIRPa association with CD47 inhibits phagocytosis of CD47 presenting cells. Studies suggest that SIRPa ligation prevents phagocytosis by inactivating paxillin and cofilin, proteins involved in cytoskeleton remodeling.