Please login to your online account to display your discounted pricing

CD172a (SIRP alpha) Mouse anti-Human, FITC, Clone: 15-414, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD172a (SIRP alpha)
Clone 15-414
Host Species Murine
Gene Alias SIRPa, SHPS-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122101

 
life technologies
LS11172941
25 Tests Each for $158.05

501122102

 
life technologies
LS11172942
100 Tests Each for $332.05
Description & Specifications

Specifications

Antigen CD172a (SIRP alpha)
Clone 15-414
Host Species Murine
Gene Alias SIRPa, SHPS-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This 15-414 monoclonal antibody reacts with human CD172a, also known as signal regulatory protein alpha (SIRP alpha). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, granulocytes, dendritic cells, and CD34+CD38- hematopoietic progenitor cells but not on T and B lymphocytes. Moreover, neurons have also been shown to express CD172. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.