Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

CD171 Mouse anti-Human, Biotin, Clone: eBio5G3 (5G3), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $289.08

Specifications

Antigen CD171
Clone eBio5G3 (5G3)
Host Species Murine
Gene Alias Neural Cell Adhesion Molecule, L1, NCAM L1
Applications Flow Cytometry,Immunohistochemistry (Frozen),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
5011790 Life Technologies
LS13171980
25μg Each for $130.68
5011791 Life Technologies
LS13171982
100μg Each for $289.08
Specifications

Description

The monoclonal antibody eBio5G3 recognizes CD171 also known as neural cell adhesion molecule L1. CD171 is a member of the Ig superfamily containing 6 extracellular Ig domains and five fibronectin type III-like repeats. CD171 has been shown to function as a cell adhesion molecule mediating homotypic and heterotypic cell-cell interactions in neuronal myelination, neurite outgrowth and regeneration. Expression of CD171 has been found on monocytes and mature monocytic-derived and follicular DCs, a minor subset of lymphocytes in addition to that found on neuronal tissue and some tumor cells lines. Expression of CD171 on tumors is thought to contribute to tumor progression. Epitope of eBio5G3 is in amino-terminal Ig-like domain.
Description & Specifications

Specifications

CD171
Murine
Flow Cytometry,Immunohistochemistry (Frozen),Western Blot
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBio5G3 (5G3)
Neural Cell Adhesion Molecule, L1, NCAM L1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer