Please login to your online account to display your discounted pricing

CD171 Mouse anti-Human, APC, Clone: 5G3, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $172.55

Specifications

Antigen CD171
Clone eBio5G3 (5G3)
Host Species Mouse
Gene Alias Neural Cell Adhesion Molecule, L1, NCAM L1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123090

 
life technologies
LS17171941
25 tests Each for $172.55
Description & Specifications

Specifications

Antigen CD171
Clone eBio5G3 (5G3)
Host Species Mouse
Gene Alias Neural Cell Adhesion Molecule, L1, NCAM L1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The monoclonal antibody eBio5G3 recognizes CD171 also known as neural cell adhesion molecule L1. CD171 is a member of the Ig superfamily containing 6 extracellular Ig domains and five fibronectin type III-like repeats. CD171 has been shown to function as a cell adhesion molecule mediating homotypic and heterotypic cell-cell interactions in neuronal myelination, neurite outgrowth and regeneration. Expression of CD171 has been found on monocytes and mature monocytic-derived and follicular DCs, a minor subset of lymphocytes in addition to that found on neuronal tissue and some tumor cells lines. Expression of CD171 on tumors is thought to contribute to tumor progression. Epitope of eBio5G3 is in amino-terminal Ig-like domain.