Please login to your online account to display your discounted pricing

CD166 (ALCAM) Rat anti-Mouse, FITC, Clone: eBioALC48, eBioscience™

Rat Monoclonal Antibody

$158.05 - $303.05

Specifications

Antigen CD166 (ALCAM)
Clone eBioALC48
Host Species Rat
Gene Alias Activated Leucocyte Cell Adhesion Molecule
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122100

 
life technologies
LS11166182
100μg Each for $303.05

501129242

 
life technologies
LS11166180
25μg Each for $158.05
Description & Specifications

Specifications

Antigen CD166 (ALCAM)
Clone eBioALC48
Host Species Rat
Gene Alias Activated Leucocyte Cell Adhesion Molecule
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

Activated leukocyte cell adhesion molecule (ALCAM; CD166) is a 105 kD cell-surface adhesion molecule belonging to the Ig superfamily of receptors. ALCAM is a type I transmembrane glycoprotein, possessing five extracellular Ig domains, a transmembrane domain, and a short cytoplasmic tail. ALCAM is involved in homotypic adhesion, as well as heterotypic adhesion with its natural ligand, CD6. A soluble isoform of ALCAM has been shown to inhibit homophilic adhesive interactions. ALCAM is expressed on a variety of cell types, including activated T and B cells, monocyte-derived dendritic cells, and thymic epithelial cells. ALCAM expression has also been used as a marker for the isolation of pluripotent stem cells.