Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD163 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD163 Ab-1, Mouse Monoclonal Antibody.

$263.49 - $1,107.14

Specifications

Antigen CD163 Ab-1
Clone 10D6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1103R7

 
thermo scientific™ lab vision™
MS1103R7
7mL Each for $705.80

MS1103S

 
thermo scientific™ lab vision™
MS1103S
1mL Each for $1,107.14

MS1103S0

 
thermo scientific™ lab vision™
MS1103S0
100µL Each for $263.49

MS1103S1

 
thermo scientific™ lab vision™
MS1103S1
500µL Each for $743.18
Description & Specifications

Specifications

Antigen CD163 Ab-1
Clone 10D6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD163 Ab-1, Mouse Monoclonal Antibody.

CD163 antigen is a type I membrane protein also known as M130 antigen, Ber-Mac3, Ki-M8 or SM4. CD163 is restricted in its expression to the monocytic/macrophage lineage. It is present on all circulating monocytes and most tissue macrophages except those found in the mantle zone and germinal centers of lymphoid follicles, interdigitating reticulum cells and Langerhan's cells.

Host Species: Mouse

Clone: 10D6

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Domains 1 to 4 of the N-terminal region

Immunogen: Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO