missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD162 (PSGL-1) Mouse anti-Human, PerCP-eFluor 710, Clone: FLEG, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $381.48

Specifications

Antigen CD162 (PSGL-1)
Clone FLEG
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245881 Life Technologies
LS46162941
25 Tests Each for $143.88
50245882 Life Technologies
LS46162942
100 Tests Each for $381.48
Specifications

Description

This FLEG monoclonal antibody recognizes human CD162, which is also known as P-selectin glyocoprotein ligand-1 (PSGL-1). This 120kDa protein exists as a homodimer on the surface of monocytes, neutrophils, granulocytes, peripheral T cells, some B cells, and a subset of CD34+ hematopoietic progenitor cells (HPCs) in the bone marrow. CD162 binds CD62P (P-selectin), CD62E (E-selectin), and CD62L (L-selectin) to mediate leukocyte interactions with each other as well as with activated platelets and endothelium. Thus, CD162 plays a critical role in leukocyte adhesion and rolling. Furthermore, the binding of CD162 to P-selectin on HPCs has been demonstrated to inhibit hematopoiesis. Studies have also demonstrated CD162 as the receptor for enterovirus 71.
Description & Specifications

Specifications

CD162 (PSGL-1)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FLEG
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test