Please login to your online account to display your discounted pricing

CD162 (PSGL-1) Mouse anti-Human, PE, Clone: FLEG, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen CD162 (PSGL-1)
Clone FLEG
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122278

 
life technologies
LS12162941
25 Tests Each for $129.05

501122279

 
life technologies
LS12162942
100 Tests Each for $332.05
Description & Specifications

Specifications

Antigen CD162 (PSGL-1)
Clone FLEG
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This FLEG monoclonal antibody recognizes human CD162, which is also known as P-selectin glyocoprotein ligand-1 (PSGL-1). This 120kDa protein exists as a homodimer on the surface of monocytes, neutrophils, granulocytes, peripheral T cells, some B cells, and a subset of CD34+ hematopoietic progenitor cells (HPCs) in the bone marrow. CD162 binds CD62P (P-selectin), CD62E (E-selectin), and CD62L (L-selectin) to mediate leukocyte interactions with each other as well as with activated platelets and endothelium. Thus, CD162 plays a critical role in leukocyte adhesion and rolling. Furthermore, the binding of CD162 to P-selectin on HPCs has been demonstrated to inhibit hematopoiesis. Studies have also demonstrated CD162 as the receptor for enterovirus 71.