missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD161 Mouse anti-Human, PE, Clone: HP-3G10, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $302.28

Specifications

Antigen CD161
Clone HP-3G10
Host Species Murine
Gene Alias NKR-P1A, NKRP1A
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010675 Life Technologies
LS12161941
25 Tests Each for $157.08
5010676 Life Technologies
LS12161942
100 Tests Each for $302.28
Specifications

Description

This HP-3G10 monoclonal antibody reacts with CD161 (also known as NKR-P1A), a member of the C-type lectin superfamily. The human homologue of NK1.1 in C57BL/6 mice, CD161 is expressed on natural killer cells and is upregulated in response to IL-12. CD161 can also be detected on various T cell subsets, including memory/effector CD4+ T cells, CD8+ T cells, gamma delta TCR T cells, and a subset of CD3+ thymocytes. Finally, CD161 expression has been demonstrated on human Th17 CD4+ T cells. The function of this receptor is unclear although studies suggest a possible stimulatory role. Nevertheless, Lectin-like transcript-1 (LLT1), which is also known as osteoclast inhibitory lectin, has been identified as the ligand for CD161.
Description & Specifications

Specifications

CD161
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test
HP-3G10
NKR-P1A, NKRP1A
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer