Please login to your online account to display your discounted pricing

CD161 Mouse anti-Human, eFluor 450, Clone: HP-3G10, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $442.25

Specifications

Antigen CD161
Clone HP-3G10
Host Species Mouse
Gene Alias NKRP1A
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50246006

 
life technologies
LS48161941
25 tests Each for $176.90

50246007

 
life technologies
LS48161942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD161
Clone HP-3G10
Host Species Mouse
Gene Alias NKRP1A
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This HP-3G10 monoclonal antibody reacts with CD161 (also known as NKR-P1A), a member of the C-type lectin superfamily. The human homologue of NK1.1 in C57BL/6 mice, CD161 is expressed on natural killer cells and is upregulated in response to IL-12. CD161 can also be detected on various T cell subsets, including memory/effector CD4+ T cells, CD8+ T cells, gamma delta TCR T cells, and a subset of CD3+ thymocytes. Finally, CD161 expression has been demonstrated on human Th17 CD4+ T cells. The function of this receptor is unclear although studies suggest a possible stimulatory role. Nevertheless, Lectin-like transcript-1 (LLT1), which is also known as osteoclast inhibitory lectin, has been identified as the ligand for CD161.