missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD160 Mouse anti-Human, PE, Clone: BY55, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD160
Clone BY55
Host Species Murine
Gene Alias BY55
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122277 Life Technologies
LS12160942
100 Tests Each for $394.68
501122276 Life Technologies
LS12160941
25 Tests Each for $157.08
Specifications

Description

This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells.
Description & Specifications

Specifications

CD160
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
BY55
BY55
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer