Learn More
CD160 Monoclonal Antibody (BY55), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 50160942
Description
Description: This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells. Applications Reported: This BY55 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BY55 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for Flow Cytometry.Specifications
CD160 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
O95971 | |
CD160 | |
Affinity chromatography | |
RUO | |
11126 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
BY55 | |
eFluor 660 | |
CD160 | |
AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |
For Research Use Only.