Please login to your online account to display your discounted pricing

CD160 Mouse anti-Human, Alexa Fluor 488, Clone: BY55, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD160
Clone BY55
Host Species Mouse
Gene Alias BY55
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50246071

 
life technologies
LS53160941
25 tests Each for $172.55

50246072

 
life technologies
LS53160942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD160
Clone BY55
Host Species Mouse
Gene Alias BY55
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma/delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells.