Please login to your online account to display your discounted pricing

CD16 Mouse anti-Human, eVolve 605, Clone: eBioCB16 (CB16), eBioscience™

Mouse Monoclonal Antibody

$201.55 - $419.05

Specifications

Antigen CD16
Clone eBioCB16 (CB16)
Host Species Murine
Gene Alias Low Affinity IgG Receptor 3, FCGR3a, IGFR4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125007

 
life technologies
LS83016841
25 Tests Each for $201.55

501125008

 
life technologies
LS83016842
100 Tests Each for $419.05
Description & Specifications

Specifications

Antigen CD16
Clone eBioCB16 (CB16)
Host Species Murine
Gene Alias Low Affinity IgG Receptor 3, FCGR3a, IGFR4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 605
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The eBioCB16 monoclonal antibody recognizes CD16 (Fc gamma RIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80 kDa. CD16 is represented by two similar genes, CD16A (Fc gamma RIIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (Fc gamma RIIIB), which exists as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1. CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3 isotypes and minimal binding of IgG2 and IgG4. Upon IgG binding, both CD16 isoforms initiate signal transduction cascades that lead to a variety of responses including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, degranulation, and proliferation.