Please login to your online account to display your discounted pricing

CD16 Mouse anti-Human, Clone: B73.1, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$317.55 - $564.05

Specifications

Antigen CD16
Clone B73.1
Host Species Mouse
Gene Alias Low Affinity IgG Receptor 3, FCGR3a, FCGR3, IGFR3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122939

 
life technologies
LS16016782
100μg Each for $317.55

501128688

 
life technologies
LS16016785
500μg Each for $564.05
Description & Specifications

Specifications

Antigen CD16
Clone B73.1
Host Species Mouse
Gene Alias Low Affinity IgG Receptor 3, FCGR3a, FCGR3, IGFR3
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

This B73.1 monoclonal antibody reacts with human CD16, a member of the Fc receptor family also known as Fc gamma RIIIa and Fc gamma RIIIb. This 50-65kDa receptor is expressed on natural killer cells, neutrophils, monocytes, and macrophages. Crosslinking of CD16 in natural killer cells and monocytes results in activation of the MAPK and PI3K signaling pathways, thus regulating the production of TNF alpha, IL-6, and IL-1 beta.

This monoclonal antibody has been shown to block Fc receptor functions.