missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD16/CD32 Rat anti-Mouse, FITC, Clone: 93, eBioscience™

Rat Monoclonal Antibody

$104.28 - $368.28

Specifications

Antigen CD16/CD32
Clone 93
Host Species Rat
Gene Alias FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122042 Life Technologies
LS11016181
50μg Each for $104.28
501122043 Life Technologies
LS11016185
500μg Each for $368.28
509459 Life Technologies
LS11016182
100μg Each for $196.68
Specifications

Description

The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils.
Description & Specifications

Specifications

CD16/CD32
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
93
FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL