missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD16/CD32 Rat anti-Mouse, eFluor 450, Clone: 93, eBioscience™

Rat Monoclonal Antibody

$143.88 - $308.88

Specifications

Antigen CD16/CD32
Clone 93
Host Species Rat
Gene Alias FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016262 Life Technologies
LS48016180
25μg Each for $143.88
5016263 Life Technologies
LS48016182
100μg Each for $308.88
Specifications

Description

The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils.
Description & Specifications

Specifications

CD16/CD32
Rat
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
93
FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer