Please login to your online account to display your discounted pricing

CD16/CD32 Rat anti-Mouse, Biotin, Clone: 93, eBioscience™

Rat Monoclonal Antibody

$114.55 - $680.05

Specifications

Antigen CD16/CD32
Clone 93
Host Species Rat
Gene Alias FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011447

 
affymetrix
13016181
50μg Each for $114.55

5011448

 
affymetrix
13016182
100μg Each for $216.05

5011449

 
affymetrix
13016185
500μg Each for $375.55

5011450

 
affymetrix
13016186
1mg Each for $680.05
Description & Specifications

Specifications

Antigen CD16/CD32
Clone 93
Host Species Rat
Gene Alias FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils.