missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD158f Mouse anti-Human, PE-Cy7, Clone: HP-MA4, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $368.28

Specifications

Antigen CD158f
Clone HP-MA4
Host Species Murine
Gene Alias KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245737 Life Technologies
LS25158941
25 Tests Each for $143.88
50245738 Life Technologies
LS25158942
100 Tests Each for $368.28
Specifications

Description

The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013.
Description & Specifications

Specifications

CD158f
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HP-MA4
KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test