Please login to your online account to display your discounted pricing

CD158f Mouse anti-Human, PE, Clone: HP-MA4, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD158f
Clone HP-MA4
Host Species Murine
Gene Alias KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010671

 
life technologies
LS12158941
25 Tests Each for $143.55

5010672

 
life technologies
LS12158942
100 Tests Each for $361.05
Description & Specifications

Specifications

Antigen CD158f
Clone HP-MA4
Host Species Murine
Gene Alias KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013.