missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD158a/h/g Mouse anti-Human, APC, Clone: HP-MA4, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $328.68

Specifications

Antigen CD158a/h/g
Clone HP-MA4
Host Species Murine
Gene Alias KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015103 Life Technologies
LS17158941
25 Tests Each for $143.88
5015104 Life Technologies
LS17158942
100 Tests Each for $328.68
Specifications

Description

The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013.
Description & Specifications

Specifications

CD158a/h/g
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HP-MA4
KIR2DL1, KIR2DS1, KIR2DS3, KIR2DS5
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test