missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CD155/PVR (Poliovirus Receptor) Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunofluorescence and flow cytometry applications.
$947.98 - $1530.23
Specifications
Antigen | CD155 Ab-1 |
---|---|
Clone | D171 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Form | Concentrated |
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
MS465B1
|
Thermo Fisher Scientific
MS465B1 |
Biotin | 200μg/mL | 500 μL |
Each for $1,232.08
|
|
|||
MS465P1
|
Thermo Fisher Scientific
MS465P1 |
Unconjugated | 200μg/mL | 500 μL |
Each for $947.98
|
|
|||
MS465PABX
|
Thermo Fisher Scientific
MS465PABX |
Unconjugated | 1mg/mL | 200 μL |
Each for $1,530.23
|
|
|||
Description
PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from a single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or α and κ,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbones, respectively. Two other splice variants, Β and γ, encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues.Host Species: Mouse
Clone: D171
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with pig, dog, rabbit, rat, hamster and mouse. Others not known.
Epitope: aa 35-50
Immunogen: HeLa cells
Molecular Weight: 60-80kDa
Positive Control: HeLa, SV 80, HEp-2, RD, A431, HL-60, U-937 cells; Monkey Vero, GMK, or TC-7 cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunofluorescence
- Flow Cytometry
- Inhibits Replication of Poliovirus in HeLa Cells
Note:
USA: RUO; Int'l: RUO
Specifications
CD155 Ab-1 | |
Immunofluorescence | |
Concentrated | |
RUO | |
Human, Non-human Primate | |
IgG1 |
D171 | |
Monoclonal | |
Mouse | |
Cell Adhesion | |
HeLa cells | |
Primary |