Please login to your online account to display your discounted pricing

CD155 Mouse anti-Human, PerCP-eFluor 710, Clone: 2H7CD155, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $530.70

Specifications

Antigen CD155
Clone 2H7CD155
Host Species Mouse
Gene Alias PVR, Polio Virus Receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123781

 
life technologies
LS46155041
25 tests Each for $220.40

501129165

 
life technologies
LS46155042
100 tests Each for $530.70
Description & Specifications

Specifications

Antigen CD155
Clone 2H7CD155
Host Species Mouse
Gene Alias PVR, Polio Virus Receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The monoclonal antibody 2H7CD155 recognizes CD155 also known as the poliovirus receptor. The CD155 protein is a member of the Ig superfamily in which 4 isoforms exist; only the alpha and delta forms are expressed on the membrane of monocytes, prostate cancer cells, as well as neurectodermal and monocytic tumor cells lines. In addition, CD155 is found during mid-gastrulation during embryonic development. CD155 is thought to play a role in adhesion by interaction with the ECM component vitronectin as well as a role in NK killing of tumor cells. CD226 and CD96 are ligands for CD155. Blocking of CD155 is under therapeutic investigation for prevention of variants of poliovirus.