missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Mouse anti-Human, PE-Cy5, Clone: 24-31, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $460.68

Specifications

Antigen CD154 (CD40 Ligand)
Clone 24-31
Host Species Murine
Gene Alias CD40L
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122925 Life Technologies
LS15154841
25 Tests Each for $196.68
501122926 Life Technologies
LS15154842
100 Tests Each for $460.68
Specifications

Description

The 24-31 monoclonal antibody reacts with human CD154, a 39kDa transmembrane glycoprotein also known as gp39 and CD40 ligand (CD40L). CD154 is a member of the TNF superfamily and is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells including B cells, monocytes/macrophages and dendritic cells, CD154 serves a crucial function in T-APC cognate interaction. CD154 interaction with CD40 transduces signals for T-dependent B-cell activation and induces B cell cycle entry. 24-31 cross-reacts with the cynomolgus monkey CD154.
Description & Specifications

Specifications

CD154 (CD40 Ligand)
Murine
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
24-31
CD40L
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer