Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Mouse anti-Human, APC-eFluor 780, Clone: 24-31, eBioscience™

Mouse Monoclonal Antibody

$230.55 - $433.55

Specifications

Antigen CD154 (CD40 Ligand)
Clone 24-31
Host Species Murine
Gene Alias CD40L
Species Reactivity Cynomolgus, Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016191

 
life technologies
LS47154841
25 Tests Each for $230.55

5016192

 
life technologies
LS47154842
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen CD154 (CD40 Ligand)
Clone 24-31
Host Species Murine
Gene Alias CD40L
Species Reactivity Cynomolgus, Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The 24-31 monoclonal antibody reacts with human CD154, a 39kDa transmembrane glycoprotein also known as gp39 and CD40 ligand (CD40L). CD154 is a member of the TNF superfamily and is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells including B cells, monocytes/macrophages and dendritic cells, CD154 serves a crucial function in T-APC cognate interaction. CD154 interaction with CD40 transduces signals for T-dependent B-cell activation and induces B cell cycle entry. 24-31 cross-reacts with the Cynomolgus monkey CD154.