Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Armenian Hamster anti-Mouse, PE-Cy7, Clone: MR1, eBioscience™

Armenian Hamster Monoclonal Antibody

$176.90 - $382.80

Specifications

Antigen CD154 (CD40 Ligand)
Clone MR1
Host Species Armenian Hamster
Gene Alias CD40L
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123339

 
life technologies
LS25154180
25μg Each for $176.90

501123340

 
life technologies
LS25154182
100μg Each for $382.80
Description & Specifications

Specifications

Antigen CD154 (CD40 Ligand)
Clone MR1
Host Species Armenian Hamster
Gene Alias CD40L
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The MR1 monoclonal antibody reacts with mouse CD154, a 39kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle.

For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1.