missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Armenian Hamster anti-Mouse, eFluor 450, Clone: MR1, eBioscience™

Armenian Hamster Monoclonal Antibody

$130.68 - $315.48

Specifications

Antigen CD154 (CD40 Ligand)
Clone MR1
Host Species Armenian Hamster
Gene Alias CD40L
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124146 Life Technologies
LS48154180
25μg Each for $130.68
501124147 Life Technologies
LS48154182
100μg Each for $315.48
Specifications

Description

The MR1 monoclonal antibody reacts with mouse CD154, a 39kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle.

For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1.
Description & Specifications

Specifications

CD154 (CD40 Ligand)
Armenian Hamster
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
MR1
CD40L
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer