missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Armenian Hamster anti-Mouse, Clone: MR1, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$104.28 - $394.68

Specifications

Antigen CD154 (CD40 Ligand)
Clone MR1
Host Species Armenian Hamster
Gene Alias CD40L
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014482 Life Technologies
LS16154181
50μg Each for $104.28
5014483 Life Technologies
LS16154182
100μg Each for $196.68
5014484 Life Technologies
LS16154185
500μg Each for $394.68
Specifications

Description

The MR1 monoclonal antibody reacts with mouse CD154, a 39kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle.

For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1.
Description & Specifications

Specifications

CD154 (CD40 Ligand)
Armenian Hamster
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
MR1
CD40L
RUO
Purified
Primary
aqueous buffer, no sodium azide