missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD153 Rat anti-Mouse, PE, Clone: RM153, eBioscience™

Rat Monoclonal Antibody

$117.48 - $341.88

Specifications

Antigen CD153
Clone RM153
Host Species Rat
Gene Alias CD30 Ligand, TNFSF8
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010658 Life Technologies
LS12153181
50μg Each for $117.48
5010659 Life Technologies
LS12153182
100μg Each for $249.48
5010660 Life Technologies
LS12153183
200μg Each for $341.88
Specifications

Description

The RM153 monoclonal antibody reacts with mouse CD153 (CD30 ligand). This 40kDa cell surface receptor, a type II transmembrane protein, is a member of the TNF superfamily. Interaction of CD30 with CD153 plays an important role in T cell activation/costimulation. Expression of CD153 on T cells does not appear to require costimulation through CD28 and is upregulated via TCR engagement and peaks 24-48 hours after activation. Signaling through CD153 is important in costimulation of pre-activated T cells.
Description & Specifications

Specifications

CD153
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
RM153
CD30 Ligand, TNFSF8
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer