missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD153 Rat anti-Mouse, Clone: RM153, eBioscience™

Rat Monoclonal Antibody

$172.55 - $332.05

Specifications

Antigen CD153
Clone RM153
Host Species Rat
Gene Alias CD30 Ligand, TNFSF8
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012922 Life Technologies
LS14153185
500μg Each for $332.05
5012921 Life Technologies
LS14153182
100μg Each for $172.55
Specifications

Description

The RM153 monoclonal antibody reacts with mouse CD153 (CD30 ligand). This 40kDa cell surface receptor, a type II transmembrane protein, is a member of the TNF superfamily. Interaction of CD30 with CD153 plays an important role in T cell activation/costimulation. Expression of CD153 on T cells does not appear to require costimulation through CD28 and is upregulated via TCR engagement and peaks 24-48 hours after activation. Signaling through CD153 is important in costimulation of pre-activated T cells.
Description & Specifications

Specifications

CD153
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RM153
CD30 Ligand, TNFSF8
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen),Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL