missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD152 (CTLA-4) Mouse anti-Human, PE, Clone: 20A, A3, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $341.88

Specifications

Antigen CD152 (CTLA-4)
Clone A3)
Host Species Murine
Gene Alias CTLA4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010654 Life Technologies
LS12152841
25 Tests Each for $130.68
5010655 Life Technologies
LS12152842
100 Tests Each for $341.88
Specifications

Description

The eBio20A monoclonal antibody reacts with human CD152, also known as cytotoxic T lymphocyte antigen-4 (CTLA-4). CTLA-4, a protein with structural similarities to CD28, is expressed on activated T cells (activated B cells may also express CTLA-4) and binds the B7 family members, CD80 (B7-1) and CD86 (B7-2), with higher affinity than CD28 does. CTLA-4 and CD28 appear to deliver opposing signals to T cells: while CD28 is a potent costimulator, CTLA-4 restricts the progression of T cells to an activated state by inhibiting IL-2 secretion and cellular proliferation. The cytoplasmic portion of CTLA-4 contains ER retention motifs, resulting in intracellular localization of a large proportion of newly synthesized CTLA-4 in response to TCR signaling.
Description & Specifications

Specifications

CD152 (CTLA-4)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
A3)
CTLA4
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test