missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD152 (CTLA-4) Mouse anti-Human, PE, Clone: 14D3, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $322.08

Specifications

Antigen CD152 (CTLA-4)
Clone 14D3
Host Species Murine
Species Reactivity Human, Rhesus Ma
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010656 Life Technologies
LS12152941
25 Tests Each for $130.68
5010657 Life Technologies
LS12152942
100 Tests Each for $322.08
Specifications

Description

The 14D3 monoclonal antibody reacts with human CD152, also known as cytotoxic T lymphocyte antigen-4 (CTLA-4). CTLA-4, a protein with structural similarities to CD28, is expressed on activated T cells (activated B cells may also express CTLA-4) and binds the B7 family members, CD80 (B7-1) and CD86 (B7-2), with higher affinity than CD28 does. CTLA-4 and CD28 appear to deliver opposing signals to T cells: while CD28 is a potent costimulator, CTLA-4 restricts the progression of T cells to an activated state by inhibiting IL-2 secretion and cellular proliferation. The cytoplasmic portion of CTLA-4 contains ER retention motifs, resulting in intracellular localization of a large proportion of newly synthesized CTLA-4 in response to TCR signaling.

The 14D3 antibody also recognizes rhesus monkey and has inhibitor activity.
Description & Specifications

Specifications

CD152 (CTLA-4)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.03μg)/test
14D3
Human, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer