Please login to your online account to display your discounted pricing

CD152 (CTLA-4) Mouse anti-Human, eFluor 660, Clone: 14D3, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD152 (CTLA-4)
Clone 14D3
Host Species Mouse
Species Reactivity Human, Rhesus Ma
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124308

 
life technologies
LS50152941
25 tests Each for $158.05

501124309

 
life technologies
LS50152942
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen CD152 (CTLA-4)
Clone 14D3
Host Species Mouse
Species Reactivity Human, Rhesus Ma
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The 14D3 monoclonal antibody reacts with human CD152, also known as cytotoxic T lymphocyte antigen-4 (CTLA-4). CTLA-4, a protein with structural similarities to CD28, is expressed on activated T cells (activated B cells may also express CTLA-4) and binds the B7 family members, CD80 (B7-1) and CD86 (B7-2), with higher affinity than CD28 does. CTLA-4 and CD28 appear to deliver opposing signals to T cells: while CD28 is a potent costimulator, CTLA-4 restricts the progression of T cells to an activated state by inhibiting IL-2 secretion and cellular proliferation. The cytoplasmic portion of CTLA-4 contains ER retention motifs, resulting in intracellular localization of a large proportion of newly synthesized CTLA-4 in response to TCR signaling.

The 14D3 antibody also recognizes rhesus monkey and has inhibitor activity.