Please login to your online account to display your discounted pricing

CD150 Rat anti-Mouse, PE, Clone: 9D1, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD150
Clone 9D1
Host Species Rat
Gene Alias SLAM, IPO-3, IPO3, SLAMF1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010642

 
life technologies
LS12150180
25μg Each for $143.55

5010643

 
life technologies
LS12150182
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD150
Clone 9D1
Host Species Rat
Gene Alias SLAM, IPO-3, IPO3, SLAMF1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 9D1 monoclonal antibody reacts with mouse CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T (especially TH1) and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. 9D1 is reported to be an activating antibody.

Mouse hematopoietic stem cells (HSC) can be identified using SLAM family markers, such as CD150+CD244-CD48-. For this application we recommend the use of antibody clone mShad150 (cat. 12-1502).