missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD150 Rat anti-Mouse, FITC, Clone: mShad150, eBioscience™

Rat Monoclonal Antibody

$117.48 - $275.88

Specifications

Antigen CD150
Clone mShad150
Host Species Rat
Gene Alias SLAM, Signaling lymphocyte activation molecule
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122096 Life Technologies
LS11150280
25μg Each for $117.48
501129149 Life Technologies
LS11150282
100μg Each for $275.88
Specifications

Description

The mShad150 monoclonal antibody reacts with mouse CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells.
Description & Specifications

Specifications

CD150
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
mShad150
SLAM, Signaling lymphocyte activation molecule
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer