missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD150 Rat anti-Mouse, FITC, Clone: 9D1, eBioscience™

Rat Monoclonal Antibody

$143.88 - $368.28

Specifications

Antigen CD150
Clone 9D1
Host Species Rat
Gene Alias SLAM, IPO-3, IPO3, SLAMF1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509772 Life Technologies
LS11150180
25μg Each for $143.88
509773 Life Technologies
LS11150182
100μg Each for $368.28
Specifications

Description

The 9D1 monoclonal antibody reacts with mouse CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T (especially TH1) and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. 9D1 is reported to be an activating antibody.

Mouse hematopoietic stem cells (HSC) can be identified using SLAM family markers, such as CD150+CD244-CD48-. For this application we recommend the use of antibody clone mShad150 (cat. 12-1502).
Description & Specifications

Specifications

CD150
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
9D1
SLAM, IPO-3, IPO3, SLAMF1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer