Please login to your online account to display your discounted pricing

CD150 Rat anti-Mouse, eFluor 450, Clone: mShad150, eBioscience™

Rat Monoclonal Antibody

$191.40 - $456.75

Specifications

Antigen CD150
Clone mShad150
Host Species Rat
Gene Alias SLAM, Signaling lymphocyte activation molecule
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124144

 
life technologies
LS48150280
25μg Each for $191.40

501124145

 
life technologies
LS48150282
100μg Each for $456.75
Description & Specifications

Specifications

Antigen CD150
Clone mShad150
Host Species Rat
Gene Alias SLAM, Signaling lymphocyte activation molecule
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The mShad150 monoclonal antibody reacts with mouse CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells.