Please login to your online account to display your discounted pricing

CD150 Mouse anti-Human, PE, Clone: A12 (7D4), eBioscience™

Mouse Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD150
Clone A12 (7D4)
Host Species Mouse
Gene Alias SLAM, IPO-3, IPO3, SLAMF1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010647

 
affymetrix
12150941
25 tests Each for $158.05

5010648

 
affymetrix
12150942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD150
Clone A12 (7D4)
Host Species Mouse
Gene Alias SLAM, IPO-3, IPO3, SLAMF1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The A12 monoclonal antibody reacts with human CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM induces proliferation and cytokine production of T cells and SLAM plays a role in activation of T and B cells.