missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD15 Mouse anti-Human, PerCP-eFluor 710, Clone: MMA, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD15
Clone MMA
Host Species Murine
Gene Alias Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245851 Life Technologies
LS46015841
25 Tests Each for $130.68
50245852 Life Technologies
LS46015842
100 Tests Each for $328.68
Specifications

Description

The MMA monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative.
Description & Specifications

Specifications

CD15
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MMA
Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test