Please login to your online account to display your discounted pricing

CD15 Mouse anti-Human, PE, Clone: HI98, eBioscience™

Mouse Monoclonal Antibody

$159.50 - $350.90

Specifications

Antigen CD15
Clone HI98
Host Species Mouse
Gene Alias Fucosyl Transferase 4 (FUT4), Alpha-3-Fucosyltransferase (FCT3A)
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122213

 
life technologies
LS12015941
25 tests Each for $159.50

501122214

 
life technologies
LS12015942
100 tests Each for $350.90
Description & Specifications

Specifications

Antigen CD15
Clone HI98
Host Species Mouse
Gene Alias Fucosyl Transferase 4 (FUT4), Alpha-3-Fucosyltransferase (FCT3A)
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative.