missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD15 Mouse anti-Human, eFluor 450, Clone: MMA, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $376.20

Specifications

Antigen CD15
Clone MMA
Host Species Murine
Gene Alias Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016258 Life Technologies
LS48015841
25 Tests Each for $183.48
5016259 Life Technologies
LS48015842
100 Tests Each for $376.20
Specifications

Description

The MMA monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative.
Description & Specifications

Specifications

CD15
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
MMA
Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer